Product Details

SNP ID
rs201108066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126304117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTCAACTAGGCAAGAGGAAG[C/T]GCGAATTGGACGTGGAGGAGGCCCA
Phenotype
MIM: 610534 MIM: 606252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 817 Missense Mutation CGC,TGC R13C NP_054745.1
XM_011542778.2 817 Missense Mutation CGC,TGC R13C XP_011541080.1
XM_011542780.2 817 Intron XP_011541082.1
XM_017017612.1 817 UTR 5 XP_016873101.1
Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details