Product Details

SNP ID
rs201362033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111879571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCTCGTCTGAAATGGCTGCCT[C/T]CCAGTGTCTCTGCTGCTCAAAATTT
Phenotype
MIM: 606941
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG9 PubMed Links

Gene Details

Gene
ALG9
Gene Name
ALG9, alpha-1,2-mannosyltransferase
There are no transcripts associated with this gene.

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022761.2 53 Missense Mutation TCC,TTC S4F NP_073598.1
XM_011542957.2 53 UTR 5 XP_011541259.1
XM_011542958.2 53 UTR 5 XP_011541260.1
XM_017018129.1 53 Missense Mutation TCC,TTC S4F XP_016873618.1
Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138378.2 53 Intron NP_612387.1

View Full Product Details