Product Details

SNP ID
rs201177986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130406125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTGACAATGTCATTGTAGCC[A/T]TAACTGTGATAGAAACAGAAGGACA
Phenotype
MIM: 605175
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS8 PubMed Links

Gene Details

Gene
ADAMTS8
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007037.5 2810 Silent Mutation TAA,TAT *701Y NP_008968.4
XM_017017145.1 2810 Silent Mutation TAA,TAT *685Y XP_016872634.1
Gene
LOC646383
Gene Name
uncharacterized LOC646383
There are no transcripts associated with this gene.

View Full Product Details