Product Details

SNP ID
rs201156914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75428466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGACGTAGACGTCATTGCGGA[A/G]AGCACAGGCGGCGAATTCTGAGCGA
Phenotype
MIM: 609632 MIM: 600454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD5 PubMed Links

Gene Details

Gene
GDPD5
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
There are no transcripts associated with this gene.

Gene
KLHL35
Gene Name
kelch like family member 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039548.2 1042 Missense Mutation CTC,TTC L348F NP_001034637.2
XM_011544954.2 1042 Missense Mutation CTC,TTC L351F XP_011543256.1
Gene
RPS3
Gene Name
ribosomal protein S3
There are no transcripts associated with this gene.

View Full Product Details