Product Details

SNP ID
rs201396149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGAACTTTGTGTCAGAGAAAA[A/T]CATCATCTCCTACGCAGGGTGCATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 320 Missense Mutation AAC,ATC N107I NP_001005194.1

View Full Product Details