Product Details

SNP ID
rs201148803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5199854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGTAGATGTTGCCAATGAGAA[C/T]GTGGGCCACGGGGGAAAGGTGCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51V1 PubMed Links

Gene Details

Gene
OR51V1
Gene Name
olfactory receptor family 51 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004760.2 847 Missense Mutation ATT,GTT I283V NP_001004760.2

View Full Product Details