Product Details

SNP ID
rs199474814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110911094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCTCCTAGTCCTTCTCTTCTC[T/C]GTGGGTGATGATGTGCACCAGGTTC
Phenotype
MIM: 160781
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CCDC63 PubMed Links

Gene Details

Gene
CCDC63
Gene Name
coiled-coil domain containing 63
There are no transcripts associated with this gene.

Gene
MYL2
Gene Name
myosin light chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000432.3 555 Missense Mutation AGA,GGA R162G NP_000423.2

View Full Product Details