Product Details

SNP ID
rs199552914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82354578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGATGTTTTCTTTTTATTTTTCT[G/T]TGGAATTGTAAAGGAGCTTTTAATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC59 PubMed Links

Gene Details

Gene
CCDC59
Gene Name
coiled-coil domain containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014167.4 525 Missense Mutation AAG,CAG K161Q NP_054886.2
Gene
METTL25
Gene Name
methyltransferase like 25
There are no transcripts associated with this gene.

View Full Product Details