Product Details

SNP ID
rs199575449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49090469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGGAAAGCAGCCGCCCGCCCGG[A/C]CCGCCAGTGAGTTATCTGCAGGGAA
Phenotype
MIM: 605423 MIM: 610007
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHH PubMed Links

Gene Details

Gene
DHH
Gene Name
desert hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021044.3 888 Missense Mutation GGC,GTC G194V NP_066382.1
XM_017019380.1 888 Missense Mutation GGC,GTC G147V XP_016874869.1
XM_017019381.1 888 Missense Mutation GGC,GTC G80V XP_016874870.1
Gene
LMBR1L
Gene Name
limb development membrane protein 1 like
There are no transcripts associated with this gene.

View Full Product Details