Product Details

SNP ID
rs201127081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132728068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGTGAGTAGCACAGGACAGCA[C/G]TCTTCCATCTGTGCACGGCCGGGAA
Phenotype
MIM: 616062 MIM: 614939
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKLE2 PubMed Links

Gene Details

Gene
ANKLE2
Gene Name
ankyrin repeat and LEM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015114.2 1988 Missense Mutation ACT,AGT T860S NP_055929.1
XM_005266159.3 1988 Missense Mutation ACT,AGT T798S XP_005266216.1
XM_005266160.1 1988 Missense Mutation ACT,AGT T798S XP_005266217.1
XM_006719735.1 1988 Missense Mutation CTG,GTG L663V XP_006719798.1
XM_011534787.2 1988 Intron XP_011533089.1
XM_011534788.1 1988 Intron XP_011533090.1
XM_017019076.1 1988 Missense Mutation ACT,AGT T423S XP_016874565.1
Gene
PGAM5
Gene Name
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
There are no transcripts associated with this gene.

View Full Product Details