Product Details

SNP ID
rs200801104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18081436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTCGCACATGACAAAGATCTC[A/G]TTTGTTGCCAACCAAAAACACTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RERGL PubMed Links

Gene Details

Gene
RERGL
Gene Name
RERG like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286201.1 580 Nonsense Mutation CGA,TGA R124* NP_001273130.1
NM_024730.3 580 Nonsense Mutation CGA,TGA R125* NP_079006.1

View Full Product Details