Product Details

SNP ID
rs200968873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55636899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCTGGCCTGGAGTGATGGCTG[C/G]GGAAAACCATACTACACTGCCTGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10P1 PubMed Links

Gene Details

Gene
OR10P1
Gene Name
olfactory receptor family 10 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206899.1 8 Missense Mutation GCG,GGG A3G NP_996782.1

View Full Product Details