Product Details

SNP ID
rs201063803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109451631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGCCCGGTCATCAGGGCGCTT[A/G]ATGTGGATCCTCCGCACGCGCTCGA
Phenotype
MIM: 613421 MIM: 614636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 1191 Silent Mutation ATC,ATT I303I NP_001304324.1
NM_001317399.1 1191 Silent Mutation ATC,ATT I279I NP_001304328.1
NM_031954.4 1191 Silent Mutation ATC,ATT I302I NP_114160.1
Gene
MYO1H
Gene Name
myosin IH
There are no transcripts associated with this gene.

View Full Product Details