Product Details

SNP ID
rs201092099
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:90954128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCTCCTGCTGACGCTCCACCTT[C/T]TCCTGCTGCCTCATGTCCTCGTAGA
Phenotype
MIM: 601657
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCER1 PubMed Links

Gene Details

Gene
CCER1
Gene Name
coiled-coil glutamate rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152638.3 1049 Silent Mutation GAA,GAG E205E NP_689851.1
Gene
EPYC
Gene Name
epiphycan
There are no transcripts associated with this gene.

Gene
LINC00615
Gene Name
long intergenic non-protein coding RNA 615
There are no transcripts associated with this gene.

View Full Product Details