Product Details

SNP ID
rs201121113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57769664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGTGGGTCGGGGAAGAGGAA[A/G]AACATCTTTGTCAGCTGTGAGACAG
Phenotype
MIM: 609506 MIM: 613336 MIM: 604466 MIM: 615258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP27B1 PubMed Links

Gene Details

Gene
CYP27B1
Gene Name
cytochrome P450 family 27 subfamily B member 1
There are no transcripts associated with this gene.

Gene
MARCH9
Gene Name
membrane associated ring-CH-type finger 9
There are no transcripts associated with this gene.

Gene
METTL1
Gene Name
methyltransferase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005371.5 640 Silent Mutation TTC,TTT F158F NP_005362.3
NM_023033.3 640 Missense Mutation CTT,TTT L97F NP_075422.3
XM_005268873.2 640 Silent Mutation TTC,TTT F115F XP_005268930.1
XM_017019305.1 640 Silent Mutation TTC,TTT F183F XP_016874794.1
Gene
METTL21B
Gene Name
methyltransferase like 21B
There are no transcripts associated with this gene.

View Full Product Details