Product Details

SNP ID
rs200791667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48922127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCCCTACCAGAGGCAAAATGC[A/C]CTTCACCAGCTTTAGCCAGTAGTTG
Phenotype
MIM: 103190 MIM: 611088 MIM: 610571
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
There are no transcripts associated with this gene.

Gene
CCDC65
Gene Name
coiled-coil domain containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286957.1 376 Intron NP_001273886.1
NM_033124.4 376 Intron NP_149115.2
Gene
FKBP11
Gene Name
FK506 binding protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143781.1 376 Missense Mutation GGC,TGC G53C NP_001137253.1
NM_001143782.1 376 Intron NP_001137254.1
NM_016594.2 376 Missense Mutation GGC,TGC G155C NP_057678.1

View Full Product Details