Product Details

SNP ID
rs201029707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98732492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACCAATCTTAAGAAAATACAC[A/G]TGTCCCCTGACTTCAAAACATATGT
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 7377 Intron NP_001190994.1
NM_001204066.1 7377 Intron NP_001190995.1
NM_001204067.1 7377 Intron NP_001190996.1
NM_001204068.1 7377 Intron NP_001190997.1
NM_001204069.1 7377 Intron NP_001190998.1
NM_001204070.1 7377 Intron NP_001190999.1
NM_001204079.1 7377 Intron NP_001191008.1
NM_001204080.1 7377 Intron NP_001191009.1
NM_001204081.1 7377 Intron NP_001191010.1
NM_020140.3 7377 Intron NP_064525.1
NM_152788.4 7377 Intron NP_690001.3
NM_181670.3 7377 Intron NP_858056.2
XM_005269028.4 7377 Intron XP_005269085.1
XM_005269029.4 7377 Intron XP_005269086.1
XM_005269032.3 7377 Intron XP_005269089.1
XM_006719504.3 7377 Intron XP_006719567.1
XM_006719505.3 7377 Intron XP_006719568.1
XM_006719506.3 7377 Intron XP_006719569.1
XM_006719507.3 7377 Intron XP_006719570.1
XM_006719508.3 7377 Intron XP_006719571.1
XM_006719509.3 7377 Intron XP_006719572.1
XM_006719510.3 7377 Intron XP_006719573.1
XM_006719512.3 7377 Intron XP_006719575.1
XM_006719513.3 7377 Intron XP_006719576.1
XM_006719514.3 7377 Intron XP_006719577.1
XM_011538571.2 7377 Intron XP_011536873.1
XM_017019651.1 7377 Intron XP_016875140.1
XM_017019652.1 7377 Intron XP_016875141.1
XM_017019653.1 7377 UTR 3 XP_016875142.1
XM_017019654.1 7377 Intron XP_016875143.1
XM_017019655.1 7377 Intron XP_016875144.1
XM_017019656.1 7377 Intron XP_016875145.1
XM_017019657.1 7377 Intron XP_016875146.1
XM_017019658.1 7377 Intron XP_016875147.1
XM_017019659.1 7377 Intron XP_016875148.1
XM_017019660.1 7377 Intron XP_016875149.1
XM_017019661.1 7377 Intron XP_016875150.1
XM_017019662.1 7377 Intron XP_016875151.1
XM_017019663.1 7377 Intron XP_016875152.1
XM_017019664.1 7377 Intron XP_016875153.1
XM_017019665.1 7377 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160.2 7377 Missense Mutation ATG,GTG M1171V NP_001151.1
NM_013229.2 7377 Missense Mutation ATG,GTG M1214V NP_037361.1
NM_181861.1 7377 Missense Mutation ATG,GTG M1225V NP_863651.1
NM_181868.1 7377 Missense Mutation ATG,GTG M1182V NP_863658.1
NM_181869.1 7377 UTR 3 NP_863659.1
XM_017019250.1 7377 Intron XP_016874739.1

View Full Product Details