Product Details

SNP ID
rs200375363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111644373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGTCTCCAGGTAGAACATGAC[A/G]TCACGCAGCTGCTCCTGGATCTCGG
Phenotype
MIM: 604986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRAP PubMed Links

Gene Details

Gene
BRAP
Gene Name
BRCA1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006768.4 1600 Silent Mutation GAC,GAT D535D NP_006759.3
XM_005253944.4 1600 Silent Mutation GAC,GAT D576D XP_005254001.1
XM_011538789.2 1600 Silent Mutation GAC,GAT D248D XP_011537091.1
XM_017019992.1 1600 Silent Mutation GAC,GAT D481D XP_016875481.1
XM_017019993.1 1600 Silent Mutation GAC,GAT D386D XP_016875482.1
XM_017019994.1 1600 Silent Mutation GAC,GAT D386D XP_016875483.1
XM_017019995.1 1600 Silent Mutation GAC,GAT D386D XP_016875484.1

View Full Product Details