Product Details

SNP ID
rs200355447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6954874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGCCAAGGGCGAGCCCTGGAC[A/G]TTTCTTGTGCGTGAGAGCCTCAGCC
Phenotype
MIM: 615140 MIM: 612093 MIM: 612092 MIM: 176883
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf57 PubMed Links

Gene Details

Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
There are no transcripts associated with this gene.

Gene
LOC105369635
Gene Name
uncharacterized LOC105369635
There are no transcripts associated with this gene.

Gene
MIR141
Gene Name
microRNA 141
There are no transcripts associated with this gene.

Gene
MIR200C
Gene Name
microRNA 200c
There are no transcripts associated with this gene.

Gene
PTPN6
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002831.5 551 Silent Mutation ACA,ACG T132T NP_002822.2
NM_080548.4 551 Silent Mutation ACA,ACG T134T NP_536858.1
NM_080549.3 551 Silent Mutation ACA,ACG T132T NP_536859.1
XM_006718994.1 551 Silent Mutation ACA,ACG T18T XP_006719057.1
XM_011520988.1 551 Silent Mutation ACA,ACG T134T XP_011519290.1

View Full Product Details