Product Details

SNP ID
rs200047743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57610275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACCGGATACTGGGGGTCATGG[C/G]GGGCATGCTGCGCGCATGCGCCCTC
Phenotype
MIM: 610215 MIM: 613142
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF25 PubMed Links

Gene Details

Gene
ARHGEF25
Gene Name
Rho guanine nucleotide exchange factor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111270.2 96 Missense Mutation GCG,GGG A20G NP_001104740.1
NM_182947.3 96 Intron NP_891992.2
Gene
DTX3
Gene Name
deltex E3 ubiquitin ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286245.1 96 Intron NP_001273174.1
NM_001286246.1 96 Intron NP_001273175.1
NM_178502.3 96 Intron NP_848597.1
XM_005268697.1 96 Intron XP_005268754.1
XM_005268698.1 96 Intron XP_005268755.1
XM_005268700.1 96 Intron XP_005268757.1
XM_005268703.1 96 Intron XP_005268760.1
XM_011538022.2 96 Intron XP_011536324.1
Gene
LOC101927583
Gene Name
uncharacterized LOC101927583
There are no transcripts associated with this gene.

Gene
PIP4K2C
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
There are no transcripts associated with this gene.

Gene
SLC26A10
Gene Name
solute carrier family 26 member 10
There are no transcripts associated with this gene.

View Full Product Details