Product Details

SNP ID
rs200165456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6905884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACGGAGGACATGATGAAGGAA[A/G]GGCTTTCTCTGCTCTGTAAGACAGG
Phenotype
MIM: 131360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO2 PubMed Links

Gene Details

Gene
ENO2
Gene Name
enolase 2
There are no transcripts associated with this gene.

Gene
LRRC23
Gene Name
leucine rich repeat containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135217.1 386 Missense Mutation AGG,GGG R56G NP_001128689.1
NM_006992.3 386 Missense Mutation AGG,GGG R56G NP_008923.1
NM_201650.2 386 Missense Mutation AGG,GGG R56G NP_964013.1

View Full Product Details