Product Details
- SNP ID
-
rs200388520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10847182 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAGAGTTGACGGTGTCCTCGTC[A/G]GGGTGGTCGTTGCTGATTTTGAAAA
- Phenotype
-
MIM: 605359
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRH1-PRR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1063193] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRH1-PRR4
- Gene Name
- PRH1-PRR4 readthrough
There are no transcripts associated with this gene.
- Gene
- PRR4
- Gene Name
- proline rich 4 (lacrimal)
View Full Product Details