Product Details

SNP ID
rs200035641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6620249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCGTCGGTGGTGACGGCGGACC[C/T]TTCGGATTGCGCGAGCGCCGCCCGC
Phenotype
MIM: 606926
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPAR5 PubMed Links

Gene Details

Gene
LPAR5
Gene Name
lysophosphatidic acid receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142961.1 1281 Missense Mutation AGG,GGG R334G NP_001136433.1
NM_020400.5 1281 Missense Mutation AGG,GGG R334G NP_065133.1

View Full Product Details