Product Details

SNP ID
rs200775835
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48202378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCCAGCCTGGTTCTTCCAAG[C/T]GCCCTCACCATGGCCTCCTTAATTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR10AD1 PubMed Links

Gene Details

Gene
OR10AD1
Gene Name
olfactory receptor family 10 subfamily AD member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004134.1 915 Silent Mutation GCA,GCG A305A NP_001004134.1

View Full Product Details