Product Details

SNP ID
rs200442958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120979025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAGACTTCACGCCACCCATCC[A/T]CAAAGAGCTGGAGAACCTCAGCCCT
Phenotype
MIM: 142410
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HNF1A PubMed Links

Gene Details

Gene
HNF1A
Gene Name
HNF1 homeobox A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000545.6 458 Missense Mutation CAC,CTC H86L NP_000536.5
NM_001306179.1 458 Missense Mutation CAC,CTC H86L NP_001293108.1
XM_005253931.3 458 Missense Mutation CAC,CTC H86L XP_005253988.1
Gene
HNF1A-AS1
Gene Name
HNF1A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details