Product Details

SNP ID
rs200445443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4370365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTAGATGAACTTGGCGAAGGGG[C/T]GGCAGCCTTCCGGGCCCGTTCCCCC
Phenotype
MIM: 605380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF23 PubMed Links

Gene Details

Gene
FGF23
Gene Name
fibroblast growth factor 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020638.2 880 Missense Mutation CAC,CGC H245R NP_065689.1

View Full Product Details