Product Details

SNP ID
rs200446908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122729512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGGGCTTGAAAAATAATACAC[C/G]AGGGGATCCAGCATGCTGTTCATGT
Phenotype
MIM: 606923
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HCAR1 PubMed Links

Gene Details

Gene
HCAR1
Gene Name
hydroxycarboxylic acid receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032554.3 1071 Silent Mutation CTC,CTG L276L NP_115943.1

View Full Product Details