Product Details

SNP ID
rs200430605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52839185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTGTGGCAGCTGGAGCCAGC[A/G]CTGGAGCCAGACACAGAGCAGGAAT
Phenotype
MIM: 611159 MIM: 611160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT78 PubMed Links

Gene Details

Gene
KRT78
Gene Name
keratin 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300814.1 1173 Silent Mutation AGC,AGT S387S NP_001287743.1
NM_173352.3 1173 Silent Mutation AGC,AGT S497S NP_775487.2
XM_011538010.1 1173 Silent Mutation AGC,AGT S465S XP_011536312.1
Gene
KRT79
Gene Name
keratin 79
There are no transcripts associated with this gene.

View Full Product Details