Product Details

SNP ID
rs200748775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130873112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAAATTCGGTGGACTGAGAGA[C/T]GGCTATTATATCCAAGGTAGGCATT
Phenotype
MIM: 601179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAN PubMed Links

Gene Details

Gene
RAN
Gene Name
RAN, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300796.1 320 UTR 5 NP_001287725.1
NM_001300797.1 320 UTR 5 NP_001287726.1
NM_006325.4 320 Silent Mutation GAC,GAT D77D NP_006316.1
XM_017019772.1 320 Silent Mutation GAC,GAT D77D XP_016875261.1
XM_017019773.1 320 UTR 5 XP_016875262.1

View Full Product Details