Product Details

SNP ID
rs200725728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14915609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTAAATTGCCAAAGCTGGCAG[C/T]TGAGACTCCTTTAGTTTGAAAAATG
Phenotype
MIM: 610642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERP27 PubMed Links

Gene Details

Gene
ERP27
Gene Name
endoplasmic reticulum protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300784.1 695 Silent Mutation CAA,CAG Q117Q NP_001287713.1
NM_152321.3 695 Silent Mutation CAA,CAG Q218Q NP_689534.1

View Full Product Details