Product Details

SNP ID
rs200774616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108591613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTGGGCTTCCTGGGCTAACAA[A/G]AGAGACCCTTCCAGCTCCCCTTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM119 PubMed Links

Gene Details

Gene
TMEM119
Gene Name
transmembrane protein 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181724.2 1124 Silent Mutation CTC,CTT L257L NP_859075.2
XM_011538271.2 1124 Silent Mutation CTC,CTT L257L XP_011536573.1

View Full Product Details