Product Details

SNP ID
rs200653575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111644298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGCCATGGCGATGTTGATCTG[C/T]CCCTCCTGGATTTCCTGCCGGGTCT
Phenotype
MIM: 604986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRAP PubMed Links

Gene Details

Gene
BRAP
Gene Name
BRCA1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006768.4 1675 Silent Mutation GGA,GGG G560G NP_006759.3
XM_005253944.4 1675 Silent Mutation GGA,GGG G601G XP_005254001.1
XM_011538789.2 1675 Silent Mutation GGA,GGG G273G XP_011537091.1
XM_017019992.1 1675 Silent Mutation GGA,GGG G506G XP_016875481.1
XM_017019993.1 1675 Silent Mutation GGA,GGG G411G XP_016875482.1
XM_017019994.1 1675 Silent Mutation GGA,GGG G411G XP_016875483.1
XM_017019995.1 1675 Silent Mutation GGA,GGG G411G XP_016875484.1

View Full Product Details