Product Details
- SNP ID
-
rs199800012
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6651209 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCCCTTATCTATTTCTTCTTCC[A/G]TTCTTGGGAGCAGCGTGGGCAAAAC
- Phenotype
-
MIM: 608352
MIM: 608524
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACRBP
PubMed Links
Gene Details
- Gene
- ACRBP
- Gene Name
- acrosin binding protein
There are no transcripts associated with this gene.
- Gene
- ING4
- Gene Name
- inhibitor of growth family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127582.1 |
774 |
Missense Mutation |
CGG,TGG |
R246W |
NP_001121054.1 |
NM_001127583.1 |
774 |
Missense Mutation |
CGG,TGG |
R243W |
NP_001121055.1 |
NM_001127584.1 |
774 |
Missense Mutation |
CGG,TGG |
R242W |
NP_001121056.1 |
NM_001127585.1 |
774 |
Missense Mutation |
CGG,TGG |
R222W |
NP_001121057.1 |
NM_001127586.1 |
774 |
UTR 3 |
|
|
NP_001121058.1 |
NM_016162.3 |
774 |
Missense Mutation |
CGG,TGG |
R245W |
NP_057246.2 |
XM_005253698.3 |
774 |
Missense Mutation |
CGG,TGG |
R197W |
XP_005253755.1 |
XM_011520964.2 |
774 |
Missense Mutation |
CGG,TGG |
R221W |
XP_011519266.1 |
XM_011520965.2 |
774 |
Missense Mutation |
CGG,TGG |
R219W |
XP_011519267.1 |
XM_017019392.1 |
774 |
Missense Mutation |
CGG,TGG |
R196W |
XP_016874881.1 |
XM_017019393.1 |
774 |
Missense Mutation |
CGG,TGG |
R193W |
XP_016874882.1 |
View Full Product Details