Product Details

SNP ID
rs199998043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112140934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCCGGACGGAACTATGTGGCA[A/G]CTGTGGTCGCAATGTCCTTGTGAAA
Phenotype
MIM: 613197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAFD1 PubMed Links

Gene Details

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143906.1 439 Missense Mutation AAC,AGC N118S NP_001137378.1
NM_006700.2 439 Missense Mutation AAC,AGC N118S NP_006691.1

View Full Product Details