Product Details

SNP ID
rs199856398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56746169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATATACCCAGAGACGATGCTTA[A/C]ATCCAAAGTCCTCTGAGGAAGAGAG
Phenotype
MIM: 606623 MIM: 176635
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B6 PubMed Links

Gene Details

Gene
HSD17B6
Gene Name
hydroxysteroid 17-beta dehydrogenase 6
There are no transcripts associated with this gene.

Gene
PRIM1
Gene Name
primase (DNA) subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000946.2 519 Missense Mutation TGT,TTT C152F NP_000937.1

View Full Product Details