Product Details

SNP ID
rs199984011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51241078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGAAGGAGGGTATGATGCAGGT[A/G]CCAGATTTGGAGCTGGGGCTACTGC
Phenotype
MIM: 607431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAZAP2 PubMed Links

Gene Details

Gene
DAZAP2
Gene Name
DAZ associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136264.1 428 Missense Mutation ACC,GCC T92A NP_001129736.1
NM_001136266.1 428 Intron NP_001129738.1
NM_001136267.1 428 Missense Mutation ACC,GCC T82A NP_001129739.1
NM_001136268.1 428 Intron NP_001129740.1
NM_001136269.1 428 Missense Mutation ACC,GCC T114A NP_001129741.1
NM_014764.3 428 Missense Mutation ACC,GCC T114A NP_055579.1
Gene
SMAGP
Gene Name
small cell adhesion glycoprotein
There are no transcripts associated with this gene.

View Full Product Details