Product Details

SNP ID
rs199672302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8518212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAGAGAGGCACAGGAGTGCACA[A/G]GTTAGAGCACCATGCAAAGCTCAAA
Phenotype
MIM: 609964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC4D PubMed Links

Gene Details

Gene
CLEC4D
Gene Name
C-type lectin domain family 4 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080387.4 363 Missense Mutation AAG,AGG K57R NP_525126.2
XM_011520632.2 363 Missense Mutation AAG,AGG K57R XP_011518934.1
XM_011520633.2 363 Missense Mutation AAG,AGG K57R XP_011518935.1

View Full Product Details