Product Details
- SNP ID
-
rs201861897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122272941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACATCTCACAGATTTCACAGTA[C/T]GGGCGTTCCTCACCCCGACTGCCAT
- Phenotype
-
MIM: 179838
MIM: 610034
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLIP1
PubMed Links
Gene Details
- Gene
- CLIP1
- Gene Name
- CAP-Gly domain containing linker protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001247997.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1417P |
NP_001234926.1 |
NM_002956.2 |
4246 |
Silent Mutation |
CCA,CCG |
P1406P |
NP_002947.1 |
NM_198240.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1371P |
NP_937883.1 |
XM_005253593.2 |
4246 |
Silent Mutation |
CCA,CCG |
P1419P |
XP_005253650.1 |
XM_006719552.3 |
4246 |
Silent Mutation |
CCA,CCG |
P1408P |
XP_006719615.1 |
XM_017019786.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1406P |
XP_016875275.1 |
XM_017019787.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1380P |
XP_016875276.1 |
XM_017019788.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1373P |
XP_016875277.1 |
XM_017019789.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1373P |
XP_016875278.1 |
XM_017019790.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1371P |
XP_016875279.1 |
XM_017019791.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1369P |
XP_016875280.1 |
XM_017019792.1 |
4246 |
Silent Mutation |
CCA,CCG |
P1334P |
XP_016875281.1 |
- Gene
- VPS33A
- Gene Name
- VPS33A, CORVET/HOPS core subunit
There are no transcripts associated with this gene.
View Full Product Details