Product Details

SNP ID
rs201886997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112022176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGTGACAAGCTGAACATGGAG[A/C]TGAGTGAGAAATACAAGCTGGACAA
Phenotype
MIM: 602287 MIM: 612755
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERP29 PubMed Links

Gene Details

Gene
ERP29
Gene Name
endoplasmic reticulum protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034025.1 428 UTR 3 NP_001029197.1
NM_006817.3 428 Missense Mutation ATG,CTG M104L NP_006808.1
XM_017018720.1 428 Intron XP_016874209.1
Gene
NAA25
Gene Name
N(alpha)-acetyltransferase 25, NatB auxiliary subunit
There are no transcripts associated with this gene.

Gene
TMEM116
Gene Name
transmembrane protein 116
There are no transcripts associated with this gene.

View Full Product Details