Product Details
- SNP ID
-
rs201583406
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56739298 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCAATGATCTGAAACATACCTTA[C/T]GGTCGGAACAGTAAATGGATCAAAC
- Phenotype
-
MIM: 176635
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRIM1
PubMed Links
Gene Details
- Gene
- PRIM1
- Gene Name
- primase (DNA) subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000946.2 |
1112 |
Missense Mutation |
ATA,GTA |
I350V |
NP_000937.1 |
View Full Product Details