Product Details

SNP ID
rs201685430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106978215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGGTGCAGCTGTCAAAAGTCTG[C/T]TAATGACCACATTTTTTAGTCCCAA
Phenotype
MIM: 616929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTERF2 PubMed Links

Gene Details

Gene
MTERF2
Gene Name
mitochondrial transcription termination factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033050.2 856 Missense Mutation AAC,AGC N167S NP_001028222.1
NM_025198.4 856 Missense Mutation AAC,AGC N167S NP_079474.2
XM_011538767.2 856 Missense Mutation AAC,AGC N167S XP_011537069.1
XM_017019985.1 856 Missense Mutation AAC,AGC N167S XP_016875474.1
XM_017019986.1 856 Missense Mutation AAC,AGC N167S XP_016875475.1
XM_017019987.1 856 Missense Mutation AAC,AGC N167S XP_016875476.1
Gene
TMEM263
Gene Name
transmembrane protein 263
There are no transcripts associated with this gene.

View Full Product Details