Product Details

SNP ID
rs201262822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52839141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGTGATGGATGTCTTCAGACTC[A/G]ACTCAACTGTCTTCTTCAGGATGGT
Phenotype
MIM: 611159 MIM: 611160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT78 PubMed Links

Gene Details

Gene
KRT78
Gene Name
keratin 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300814.1 1217 Missense Mutation TCG,TTG S402L NP_001287743.1
NM_173352.3 1217 Missense Mutation TCG,TTG S512L NP_775487.2
XM_011538010.1 1217 Missense Mutation TCG,TTG S480L XP_011536312.1
Gene
KRT79
Gene Name
keratin 79
There are no transcripts associated with this gene.

View Full Product Details