Product Details
- SNP ID
-
rs201429050
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52768887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTGCACCGCCGAGCCTGCTCCC[A/G]CTACTGCTGCTCTGGTAGCTCCCGC
- Phenotype
-
MIM: 616671
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT76
PubMed Links
Gene Details
- Gene
- KRT76
- Gene Name
- keratin 76
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015848.4 |
1797 |
Silent Mutation |
AGC,AGT |
S581S |
NP_056932.2 |
View Full Product Details