Product Details

SNP ID
rs201454069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110853406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTCCTCTCCATCTCCCCCAGT[A/T]GAAGAAGAACAGGAGAAAAGACTCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC63 PubMed Links

Gene Details

Gene
CCDC63
Gene Name
coiled-coil domain containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286243.1 148 UTR 5 NP_001273172.1
NM_001286244.1 148 Intron NP_001273173.1
NM_152591.2 148 Nonsense Mutation TAG,TTG *4L NP_689804.1
XM_006719262.1 148 Nonsense Mutation TAG,TTG *4L XP_006719325.1
XM_006719263.2 148 Nonsense Mutation TAG,TTG *4L XP_006719326.1
XM_011537999.1 148 Nonsense Mutation TAG,TTG *4L XP_011536301.1
XM_011538001.2 148 Nonsense Mutation TAG,TTG *4L XP_011536303.1

View Full Product Details