Product Details

SNP ID
rs201265123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112163160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCTCCTGGTTGCAGGCAAACTT[C/G]ATGAACTTGCACAGCTCCTCCTGGG
Phenotype
MIM: 613197
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HECTD4 PubMed Links

Gene Details

Gene
HECTD4
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109662.3 12996 Missense Mutation ATC,ATG I4200M NP_001103132.3
Gene
MIR6861
Gene Name
microRNA 6861
There are no transcripts associated with this gene.

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
There are no transcripts associated with this gene.

View Full Product Details