Product Details

SNP ID
rs201217118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:61411264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATCAAATATTAGAAATATCCAT[G/T]GGCTTTCTCTCTCGCATATGCAAGT
Phenotype
MIM: 614449
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDH20 PubMed Links

Gene Details

Gene
PCDH20
Gene Name
protocadherin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022843.3 4635 Silent Mutation CCA,CCC P945P NP_073754.2
XM_017020708.1 4635 Silent Mutation CCA,CCC P945P XP_016876197.1

View Full Product Details