Product Details

SNP ID
rs201605208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:61411246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTATTCCACCATGAAATATCA[A/G]ATATTAGAAATATCCATTGGCTTTC
Phenotype
MIM: 614449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH20 PubMed Links

Gene Details

Gene
PCDH20
Gene Name
protocadherin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022843.3 4653 Silent Mutation ATC,ATT I951I NP_073754.2
XM_017020708.1 4653 Silent Mutation ATC,ATT I951I XP_016876197.1

View Full Product Details