Product Details

SNP ID
rs201150464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:77896470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTATTGCTCTTTCTTTCTGGC[A/C]ACATTGTTGGGTTTTGGTTTACTGT
Phenotype
MIM: 131244
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EDNRB PubMed Links

Gene Details

Gene
EDNRB
Gene Name
endothelin receptor type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000115.3 3482 UTR 3 NP_000106.1
NM_001122659.2 3482 UTR 3 NP_001116131.1
NM_001201397.1 3482 UTR 3 NP_001188326.1
NM_003991.3 3482 Missense Mutation TGG,TTG W428L NP_003982.1
Gene
EDNRB-AS1
Gene Name
EDNRB antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details