Product Details

SNP ID
rs201654359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41575760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACCTTCAGAATTTCTAGTCTTT[G/T]CTTATTGTCCTTGGGGATTTTGTTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR5006 PubMed Links

Gene Details

Gene
MIR5006
Gene Name
microRNA 5006
There are no transcripts associated with this gene.

Gene
VWA8
Gene Name
von Willebrand factor A domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009814.1 5420 Intron NP_001009814.1
NM_015058.1 5420 Missense Mutation AAA,CAA K1784Q NP_055873.1
XM_006719791.3 5420 Missense Mutation AAA,CAA K1429Q XP_006719854.1
XM_011535007.2 5420 Intron XP_011533309.1
XM_017020469.1 5420 Missense Mutation AAA,CAA K1723Q XP_016875958.1
XM_017020470.1 5420 Intron XP_016875959.1
XM_017020471.1 5420 Intron XP_016875960.1
XM_017020472.1 5420 Missense Mutation AAA,CAA K1038Q XP_016875961.1
XM_017020473.1 5420 Missense Mutation AAA,CAA K1021Q XP_016875962.1
XM_017020474.1 5420 Intron XP_016875963.1

View Full Product Details