Product Details

SNP ID
rs200740227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28303335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGGCCTGCTGACATCAGACAG[A/C]CCCGACTCCTTACTTTTACTGGTTA
Phenotype
MIM: 165070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLT1 PubMed Links

Gene Details

Gene
FLT1
Gene Name
fms related tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159920.1 4134 Intron NP_001153392.1
NM_001160030.1 4134 Intron NP_001153502.1
NM_001160031.1 4134 Intron NP_001153503.1
NM_002019.4 4134 Silent Mutation NP_002010.2
XM_011535014.1 4134 Intron XP_011533316.1
XM_017020485.1 4134 Silent Mutation XP_016875974.1
Gene
PAN3
Gene Name
PAN3 poly(A) specific ribonuclease subunit
There are no transcripts associated with this gene.

View Full Product Details